Order Entry
Germany
ContactUsLinkComponent
Anti-MME Mouse Monoclonal Antibody (CF555) [clone: CB-CALLA]
Anti-MME Mouse Monoclonal Antibody (CF555) [clone: CB-CALLA]
Supplier:  Biotium
Certificates
Anti-MME Mouse Monoclonal Antibody (CF555) [clone: CB-CALLA]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC550146-500
  • BNC550146-100
  • Antigen symbol:
  • MME
  • MME
  • Antigen name:
  • CD10
  • CD10
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 555
  • CF® 555
  • Clone:
  • Clone CB-CALLA
  • Clone CB-CALLA
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 307734
  • 307734
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications