Anti-Hepatocyte Specific Antigen Mouse Monoclonal Antibody [clone: HSA133]
Supplier: Biotium
Certificates
About this item
MAb HSA133 stains human liver canaliculi and a subset of hepatocellular carcinomas. In frozen sections, it stains liver canaliculi strongly and may be used as a marker of this hepatic substructure. Cell preparations of hepatocellular carcinoma biopsies and cell lines are found to bind this MAb on the cell surface. HSA133 strongly stains liver canaliculi and hepatic carcinoma cells using frozen sections or paraformaldehyde fixed cell preparations.
- Purified
Type: Primary
Antigen: Hepatocyte Specific Antigen
Clonality: Monoclonal
Clone: HSA133
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- BNUB0133-100
- BNUB0133-500
- Antigen name:
- Hepatocyte Specific Antigen
- Hepatocyte Specific Antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone HSA133
- Clone HSA133
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide