F(ab')2 Anti-IgY Donkey Goat Polyclonal Antibody (Alexa Fluor® 488)
JACK703-546-155
- Antigen name:Immunoglobulin Y
- Antigen symbol:IgY
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Goat
- Epitope:Heavy and Light Chains
- Cross adsorption:(min X Bov,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat,Shp Sr Prot)
- Form:Freeze-dried powder
- Antibody fragment:F(ab')2
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:300 μg
- Pk:0,3 mg
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine Red-X
Epitope: Heavy and Light Chains
Host: Donkey
Isotype: IgG
Reactivity: Goat