Anti-Mouse IgG (Fcgamma Fragment) Polyclonal Antibody (Alexa Fluor® 647)
Catalog # JACK315-605-046
Supplier: Jackson Immuno Research
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Epitope:F(c) γ
- Cross adsorption:(min X Hu Sr Prot)
- Format:Freeze-dried powder
- Antibody fragment:F(c) γ
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: Fc gamma fragment
Host: Rabbit
Isotype: IgG
Reactivity: Mouse