F(ab')2 Anti-IgM µ Rabbit Goat Polyclonal Antibody (AMCA (7-amino-4-methylcoumarin-3-acetic acid))
JACK309-156-043
- Antibody type:Secondary
- Antigen name:Immunoglobulin M mu
- Antigen symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:AMCA (7-amino-4-methylcoumarin-3-acetic acid)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chicken
- Western blot:Yes
- Epitope:F(c)'5
- Cross adsorption:No
- Format:Freeze-dried powder
- Antibody fragment:F(ab')2
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0 - 1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:500 μg
- Pk:0,5 mg
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Alkaline Phosphatase
Epitope: Heavy and Light Chains
Host: Donkey
Isotype: IgG
Reactivity: Chicken IgY (IgG) (H+L)