Anti-IgG Mouse Monoclonal Antibody (Alexa Fluor® 647)
JACK211-602-171
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Monoclonal
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Epitope:Heavy and Light Chains
- Cross adsorption:(min X Bov,Gt,Arm Hms,Hrs,Hu,Ms,Rat,Shp Ig)
- Format:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,4 - 1,8 mg/ml
- Shipping temperature:ambient
- Purification:IgG Frac
- Size:500 μg
- Pk:500 µG
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope: Fc γ
Host: Sheep
Isotype: IgG
Reactivity: Mouse IgG, Fcγ Fragment Specific