F(ab')2 Anti-IgG Mouse Goat Polyclonal Antibody (Alexa Fluor® 488)
JACK205-545-108
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Epitope:Heavy and Light Chains
- Cross adsorption:(min X Hu,Ms,Rb Sr Prot)
- Form:Freeze-dried powder
- Antibody fragment:F(ab')2
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1 mg
- Pk:1 mg
Type:
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope: F(c)'5
Host: Rabbit
Isotype: IgG
Reactivity: Human