F(ab')2 Anti-IgM µ Mouse Monoclonal Antibody (Cy3®)
Catalog # JACK200-162-156
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Digoxin
- Antigen symbol:IgM µ
- Clonality:Monoclonal
- Conjugation:Cy3®
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Form:Sterile-filtered liquid
- Antibody fragment:F(ab')2
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Molecular weight:~100 kDa
- Storage temperature:+4 °C
- Concentration:1,4 - 1,8 mg/ml
- Shipping temperature:ambient
- Purification:IgG Frac
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type:
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)'5
Host: Rabbit
Isotype: IgG
Reactivity: Human