Anti-IgG Rabbit Polyclonal Antibody (Alexa Fluor® 647)
JACK200-152-156
- Antigen name:Digoxin
- Antigen symbol:IgG
- Clonality:Monoclonal
- Conjugation:AMCA (7-amino-4-methylcoumarin-3-acetic acid)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Dog
- Cross adsorption:No
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,4 - 1,8 mg/ml
- Shipping temperature:ambient
- Purification:IgG Frac
- Size:500 μg
- Pk:0,5 mg
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Dog