Anti-Digoxin Anti-Digoxin Mouse Monoclonal Antibody
Catalog # JACK200-002-156
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Digoxin
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Cross adsorption:No
- Format:IgG Frac
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6
- Storage temperature:+4 °C
- Concentration:1,2 - 1,4 mg/ml
- Shipping temperature:ambient
- Purification:IgG Frac
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type:
Antigen: Digoxin
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor®647
Epitope: Unknown
Host: Mouse
Isotype: IgG
Reactivity: