Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 488)
Catalog # JACK127-545-160
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Armenian hamster
- Epitope:Heavy and Light Chains
- Cross adsorption:(min X Bov,Hu,Ms,Rb,Rat Sr Prot)
- Format:IgG Frac
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type:
Antigen: FITC
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor® 594
Epitope: Unknown
Host: Mouse
Isotype: IgG
Reactivity: