Anti-HRP Goat Polyclonal Antibody (Alexa Fluor® 647)
JACK123-605-021
- Antigen name:Horseradish Peroxidase
- Antigen symbol:HRP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Cross adsorption:No
- Format:IgG Frac
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1,5 mg
- Pk:1,5 mg
Type:
Antigen: FITC
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: Unknown
Host: Mouse
Isotype: IgG
Reactivity: