Anti-IgG1+IgG2a+IgG2b+IgG3 Anti-IgG1+IgG2a+IgG2b+IgG3 Goat Polyclonal Antibody (Alexa Fluor® 594)
Catalog # JACK115-585-164
Supplier: Jackson Immuno Research
Specifications
- Antigen symbol:IgG1+IgG2a+IgG2b+IgG3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 594
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:Fc gamma
- Cross adsorption:(min X Hu,Bov,Rb Sr Prot)
- Format:IgG Frac
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type:
Antigen: IgG1+IgG2a+IgG2b+IgG3
Clonality: Monoclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: