Anti-IgM µ Goat Polyclonal Antibody (Rhodamine Red-X)
Catalog # JACK115-295-075
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin M mu
- Antigen symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:Rhodamine Red-X
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Cross adsorption:(min X Hu,Bov,Hrs Sr Prot)
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0 - 1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type:
Antigen: HRP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: Unknown
Host: Goat
Isotype: IgG
Reactivity: