Anti-IgG Goat Polyclonal Antibody (Cy5®)
Catalog # JACK115-175-166
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Cy5®
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Epitope:Heavy and Light Chains
- Cross adsorption:(min X Hu,Bov,Hrs,Rb,Rat Sr Prot)
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0 - 1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type:
Antigen: IgG1+IgG2a+IgG2b+IgG3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 594
Epitope: Fc γ
Host: Goat
Isotype: IgG
Reactivity: Mouse