Anti-IgM µ Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
JACK115-095-075
- Antigen name:Immunoglobulin M mu
- Antigen symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Cross adsorption:(min X Hu,Bov,Hrs Sr Prot)
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0 - 1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1 mg
- Pk:1,5 mg
Type:
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Mouse