Anti-IgG2b Goat Polyclonal Antibody (Cy3®)
Catalog # JACK115-055-207
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G2b
- Antigen symbol:IgG2b
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:F(c) γ
- Cross adsorption:(min X Hu,Bov,Rb Sr Prot)
- Form:Freeze-dried powder
- Storage buffer:0,01M Tris-HCl, 0,25M NaCl, pH 8.0
- Storage temperature:+4 °C
- Concentration:0,6 - 1,0 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:500 μL
- Pk:0,5 mL
Specifications
About this item
Type:
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Mouse