Anti-IgG3 Goat Polyclonal Antibody
JACK115-005-209
- Antigen name:Immunoglobulin G3
- Antigen symbol:IgG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:F(c) γ
- Cross adsorption:(min X Hu,Bov,Rb Sr Prot)
- ChIP:Yes
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6
- Storage temperature:+4 °C
- Concentration:1,2 - 1,4 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1 mg
- Pk:1 mg
Type:
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Biotin-SP
Epitope: Unknown
Host: Goat
Isotype: IgG
Reactivity: Mouse