Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 488)
JACK112-545-008
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:F(c) γ
- Cross adsorption:No
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1,5 mg
- Pk:1,5 mg
Type:
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope: Unknown
Host: Goat
Isotype: IgG
Reactivity: Mouse