Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 594)
Catalog # JACK112-065-175
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin-SP (long spacer)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:Light Chain
- Cross adsorption:(min X Bov,Gt,Hrs,Hu,Ms,Rb,Shp Ig)
- ChIP:Yes
- Form:Freeze-dried powder
- Antibody fragment:Fab
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0 - 2,0 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:500 μL
- Pk:500 µl
Specifications
About this item
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 594
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Rat