F(ab')2 Anti-IgA alpha Goat Polyclonal Antibody (Alexa Fluor® 647)
Catalog # JACK109-606-011
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin A alpha
- Antigen symbol:IgA alpha
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Cross adsorption:No
- Form:Freeze-dried powder
- Antibody fragment:F(ab')2
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:750 μg
- Pk:0,75 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rabbit