Order Entry
Germany
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 488)
Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 488)
Catalog # JACK109-605-098
Anti-IgG Goat Polyclonal Antibody (Alexa Fluor® 488)
Catalog # JACK109-605-098

Specifications

  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rabbit
  • Epitope:
    F(c) γ
  • Cross adsorption:
    (min X Bov,Hrs,Ms Sr Prot)
  • Form:
    Freeze-dried powder
  • Storage buffer:
    0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
  • Storage temperature:
    +4 °C
  • Concentration:
    1,0-1,5 mg/ml
  • Shipping temperature:
    ambient
  • Purification:
    Affinity Purified
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rabbit