Anti-IgG Goat Polyclonal Antibody (Gold, colloidal 4nm)
JACK109-605-097
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Epitope:F(ab')2
- Cross adsorption:(min X Bov,Hrs,Ms Sr Prot)
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:1 mg
- Pk:1 mL
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, colloidal 4nm
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Rabbit