F(ab')2 Anti-IgG Goat Polyclonal Antibody (Peroxidase)
Catalog # JACK109-036-008
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Peroxidase
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:F(c) γ
- Cross adsorption:No
- Form:Freeze-dried powder
- Antibody fragment:F(ab')2
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA
- Storage temperature:+4 °C
- Concentration:0,8 mg/ml
- Shipping temperature:ambient
- Purification:Affinity Purified
- Size:500 μL
- Pk:0,5 mL
Specifications
About this item
Type:
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine Red-X
Epitope: F(c)'5
Host: Goat
Isotype: IgG
Reactivity: Human