Anti-IgG Goat Polyclonal Antibody (Peroxidase)
Catalog # JACK017-220-003
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Cy2®
- ELISA:Yes
- Flow cytometry:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:F(c) γ
- Cross adsorption:(min X Bov,Hrs,Ms Sr Prot)
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,4 - 2,0 mg/ml
- Shipping temperature:ambient
- Purification:Chromatographically purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Human