Anti-IgY Goat Polyclonal Antibody (Alexa Fluor® 488)
JACK016-540-084
- Antigen name:Immunoglobulin Y
- Antigen symbol:IgY
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chicken
- Epitope:Heavy and Light Chains
- Cross adsorption:(min X Bov,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat,Shp Sr Prot)
- Form:Freeze-dried powder
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Storage temperature:+4 °C
- Concentration:1,0-1,5 mg/ml
- Shipping temperature:Ambient
- Purification:Affinity Purified
- Size:500 μg
- Pk:1 mg
Type:
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope: Heavy and Light Chains
Host: Goat
Isotype: IgG
Reactivity: Chicken