Anti-IgG Mouse Polyclonal Antibody
JACK012-000-007
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- ChIP:Yes
- Neutralisation:Yes
- Form:Sterile-filtered liquid
- Antibody fragment:Fab
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6
- Molecular weight:~ 50 kDa
- Storage temperature:+4 °C
- Concentration:4-5 mg/ml
- Shipping temperature:ambient
- Purification:Chromatographically purified
- Size:2 mg
- Pk:2 mg
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Unknown
Host: Mouse
Isotype: IgG
Reactivity: Mouse