Anti-IgG Sheep Polyclonal Antibody
Catalog # JACK011-160-003
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Cy3®
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Sheep
- Western blot:Yes
- ChIP:Yes
- Neutralisation:Yes
- Form:Sterile-filtered liquid
- Antibody fragment:Fc
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Molecular weight:~ 50 kDa
- Storage temperature:+4 °C
- Concentration:1,4 - 2,0 mg/ml
- Shipping temperature:ambient
- Purification:Chromatographically purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Unknown
Host: Sheep
Isotype: IgG
Reactivity: Sheep