Anti-IgG Syrian Hamster Polyclonal Antibody
Catalog # JACK005-220-003
Supplier: Jackson Immuno Research
Specifications
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Cy2®
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Syrian hamster
- Western blot:Yes
- ChIP:Yes
- Neutralisation:Yes
- Form:Sterile-filtered liquid
- Storage buffer:0,01M Sodium Phosphate, 0,25M NaCl, pH 7.6, 15mg/mL BSA, 0,05% Azide
- Molecular weight:~160 kDa
- Storage temperature:+4 °C
- Concentration:1,4 - 2,0 mg/ml
- Shipping temperature:ambient
- Purification:Chromatographically purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Unknown
Host: Syrian Hamster
Isotype: IgG
Reactivity: Syrian Hamster