Order Entry
Germany
ContactUsLinkComponent
Anti-HS1 Mouse Monoclonal Antibody [clone: 3A3]
Anti-HS1 Mouse Monoclonal Antibody [clone: 3A3]
Catalog # BMOLKAM-TK010E
Anti-HS1 Mouse Monoclonal Antibody [clone: 3A3]
Catalog # BMOLKAM-TK010E

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HS1
  • Clonality:
    Monoclonal
  • Clone:
    3A3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Storage buffer:
    PBS, pH 7,2, containing 50% glycerol. No preservatives.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Shipped on Blue Ice
  • Immunogen:
    Recombinant human HS1.
  • Purification:
    purified by Protein A affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: HS1
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat