Anti-RANGEF Mouse Monoclonal Antibody [clone: 3D11]
BMOLKAM-CC225E
- Antibody type:Primary
- Antigen name:Regulator of chromosome condensation 1
- Antigen symbol:RCC1
- Clonality:Monoclonal
- Clone:3D11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CHC1|RCC1-I
- Storage buffer:PBS, pH 7,2, containing 1% sucrose. No preservatives. For a stock solution of 1.0mg/ml, reconstitute with 100mg/ml of sterile distilled water.
- Storage temperature:–20 °C
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Recombinant human RanGEF.
- Purification:purified by Protein A affinity chromatography
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: RANGEF
Clonality: Monoclonal
Clone: 3D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat