Capillary columns, Rxi®-Volatiles (fused silica)
: Restek
Rtx®-Volatiles columns are designed specifically for the analyses of 34 volatile organic pollutants listed in U.S. EPA methods 601, 602, and 624. With these columns, all compounds can be quantified using mass spectrometer or a PID in tandem with an ELCD.
- Proprietary Crossbond® diphenyl dimethyl polysiloxane phase
- Application-specii c columns for volatile organic pollutants
- Provides low bleed and thermal stability up to 280 °C
- Narrow bore columns can interface directly in GC/MS systems