Capillary columns, MXT®-50 (Siltek®-treated stainless steel)
Supplier: Restek
Certificates
About this item
MXT®-50 columns are general purpose columns for pesticides, herbicides, rosin acids, phthalate esters and sterols.
- Midpolarity phase; Crossbond® diphenyl dimethyl polysiloxane
- Temperature range up to 300 °C
- Equivalent to USP G3 phase
Coil diameter: 114,3 mm