Bovine fibrinogen
Supplier: SEQENS
Certificates
About this item
This ready-to-use 5% frozen solution enables a direct use of the product without further manipulation.
- Total proteins: >35,0 g/L
- Shelf life: 3 years
- Sterile filtered 0,2 µm
SEQENS IVD provides highly purified fibrinogen manufactured according to Cohn fractionation method. The plasma used is collected from healthy animals of French origin. Bovine Fibrinogen is developed for RPF supplement, used with Baird-Parker agar, for the detection of Staphylococcus aureus.
Full traceability and batch-to-batch repeatability guaranteed.
Certifications: Bovine plasma used for the manufacture of this product is collected from French healthy cattle declared free from BSE disease in its clinical forms. It is sourced in EU-approved French slaughterhouses.
Delivery: Supplied as a ready to use solution in aqueous saline buffer containing trisodium citrate and sodium chloride.
Specifications
- Species:Bovine
- Protein/peptide name:Fibrinogen
- Concentration:5% frozen solution