Order Entry
Germany
ContactUsLinkComponent
Bovine fibrinogen
Bovine fibrinogen
Supplier:  SEQENS
Certificates
undefined
Bovine fibrinogen
Supplier:  SEQENS

Specifications

  • Species:
    Bovine
  • Protein/peptide name:
    Fibrinogen
  • Concentration:
    5% frozen solution

Specifications