Order Entry
Germany
ContactUsLinkComponent
Lymphocyte separation medium, LymphoSep™, MP Biomedicals
Lymphocyte separation medium, LymphoSep™, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Lymphocyte separation medium, LymphoSep™, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Catalog No:
  • ICNA091692249
  • ICNA091692254
  • Formulation:
  • Lymphosep® is supplied as a ready made sterile aqueous solution of sodium metrizoate (9,6% w/v) and Ficoll® (5,6% w/v)
  • Osmolality:
  • 260 - 310 mOsm
  • pH range:
  • 6.0 - 9.0
  • Sterility:
  • Sterile
  • Physical form:
  • Clear, Colorless Liquid

Specifications

Frequently Bought Together