HPLC columns, Viva C4 (USP L26)
: Restek
These columns are based on a wide pore material designed for optimal large-molecule separations. Base-deactivated, wide-pore packing exhibits excellent peak shape for a wide range of compounds. Less retention in reversed phase assays than Viva C18 or Viva C8.
- Rugged, spherical particles with 300 Å pore size
- High proportion of pore/surface area available to large molecules
- Excellent for separating peptides or proteins
- Catalog No:
- REST951257E
- REST9512552-700
- REST9512553-700
- REST9512552
- REST9512512-700
- REST9512555-700
- REST9512513-700
- REST9512551
- REST9512535
- REST9512572
- REST9512571
- REST9512565
- REST9512533-700
- REST9512573-700
- REST9512535-700
- REST9512562
- REST9512561
- REST9512575-700
- REST951253E
- REST9512555
- REST951250212
- REST951250210
- REST951251E
- REST9512563-700
- REST9512522-700
- REST9512565-700
- REST9512523-700
- REST9512525-700
- REST9512521
- REST951252E
- REST9512515
- REST951256E
- REST9512532-700
- REST9512575
- REST9512572-700
- REST9512512
- REST9512511
- REST9512532
- REST9512531
- REST9512525
- REST9512515-700
- REST9512522
- REST951255E
- REST9512562-700
- Carbon load:
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26