Analytical HPLC columns, Ultra Aromax (USP L11)
Supplier: Restek
Certificates
About this item
These Ultra Aromax is a unique reversed-phase material that exhibits extreme retention and selectivity for aromatic and/or unsaturated compounds. They are made of high-purity, type-B silica that minimises activity and creates high-density bonding, these columns are designed for selective and reliable HPLC applications.
- It is an alternative to Biphenyl phase when increased retention is required
- Suitable for gradient LC-MS analyses when conventional columns are not giving adequate retention or selectivity
Specifications
- Catalog No:
- REST9127552
- REST9127552-700
- REST912750210
- REST9127531
- REST9127532
- REST9127532-700
- REST9127533-700
- REST9127535
- REST9127535-700
- REST912753E
- REST9127351
- REST9127551
- REST9127365
- REST9127365-700
- REST912736E
- REST9127525
- REST9127525-700
- REST912752E
- REST9127352-700
- REST9127353-700
- REST9127355
- REST9127355-700
- REST912751E
- REST9127521
- REST912735E
- REST9127361
- REST9127522
- REST9127362-700
- REST9127522-700
- REST9127363-700
- REST9127523-700
- REST912733E
- REST9127352
- REST9127335-700
- REST9127513-700
- REST9127515
- REST9127515-700
- REST9127315
- REST9127315-700
- REST912731E
- REST9127331
- REST9127332
- REST9127511
- REST9127332-700
- REST9127512
- REST9127333-700
- REST9127335
- REST9127512-700
- REST9127575-700
- REST912757E
- REST9127562-700
- REST9127313-700
- REST9127362
- REST9127572-700
- REST9127575
- REST9127573-700
- REST9127562
- REST9127312
- REST9127312-700
- REST9127555
- REST912750212
- REST9127555-700
- REST912755E
- REST9127311
- REST9127561
- REST9127565
- REST9127563-700
- REST9127565-700
- REST912756E
- REST9127572
- REST9127571
- REST9127553-700
- Carbon load:
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- Endcapped:
- Yes, Yes
- Yes, Yes
- N/A, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- N/A, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L11, L11
- L11, L11
- L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11
- L11, L11