SMART Digest™ proteinase K kits
Thermo Scientific™ SMART Digest™ Proteinase K Kits are designed to provide a simple to use, fast, highly reproducible and robust process for digestion of proteins in a high throughput format, meeting the needs of biopharmaceutical, bioanalytical and QA/QC environments. This version of the kit utilises Proteinase K, which cleaves the peptide bond adjacent to the carboxyl gropu of aliphatic and aromatic amino acids with blocked alpha amino groups.
- Simple process with significantly fewer steps compared to in-solution digests
- Reduced sample preparation time, typically taking <60 minutes (dependant on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours
- Higher reproducibility and greater sensitivity than other methods
- Easily automated for high throughput processing of samples
: Available as a stand alone kit, or with optional filter plate or SOLAµ™ SPE plate. Bulk resin (-B) and magnetic bulk resin (-MB) are also available separately.