Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Resprep®, CarboPrep®
Solid phase extraction cartridges, Resprep®, CarboPrep®
 :   Restek
Solid phase extraction cartridges, Resprep®, CarboPrep®
 :  Restek
Solid phase extraction cartridges, Resprep®, CarboPrep®
Solid phase extraction cartridges, Resprep®, CarboPrep®
CarboPrep® cartridges are manufactured from chromatographic-grade, nonporous, graphitized carbon. Our manufacturing process minimises variability and improves recovery and cleanup procedures.
  • Improved recovery of sulfonylurea herbicides, phenols, carbamates, and triazine herbicides, compared to C18 and C8 cartridges
  • Wide range of selectivity for both analytes and their metabolites or degradation products.
  • Rapid sampling flow rates; uncompromised recoveries
  • Maximum capacity for contaminant cleanup
  • Controlled manufacturing improves cleanliness and ensures reproducible performance.
  • Excellent performance removing pigments from samples
CarboPrep® 90 has a surface area of approximately 90 m²/g, and CarboPrep® 200 has a surface area of 200 m²/g. Both have higher capacity than silica-based packings for a variety of compounds. CarboPrep® cartridges can be used for sample extraction of organic compounds and extract cleanup to remove matrix interferences, including highly pigmented materials.
Product Family Options
  • Description
    Column Format
    Capacity
    Bed weight
  • CarboPrep® 90
    Cartridge
    3 ml
    250 mg
    1 * 50 ST
    1 * 50 ST - 395,00 €
    395,00 €
    1 * 50 ST
       395,00 €
  • CarboPrep® 90
    Cartridge
    6 ml
    500 mg
    1 * 30 ST
    1 * 30 ST - 424,00 €
    424,00 €
    1 * 30 ST
       424,00 €