Order Entry
Germany
ContactUsLinkComponent
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9048-46-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    +4 °C
  • Form:
    Powder
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Purity:
    >97%
  • Grade:
    Stem cell grade

Specifications