Gel documentation system, Quantum CX5 Edge
About this item
The Quantum CX5 Edge gel documentation system is an ultra-modern, fully automated, yet compact system with innovative features and superior technology. With just one click, it allows the recording of images of the highest quality ('One-Button-Imaging').
- Easy to operate, standalone
- 'Scientific-grade' camera with 5 MP resolution and 6× automated zoom lens
- Robust and light-proof all-metal darkroom
- Including extendable transilluminator (optional as UV, SUPER-BRIGHT, blue and white light) usable with only one hand and interchangeable
- Unprecedented sensitivity on a receiving surface of up to 21×26 cm
For maximum flexibility in fluorescence applications an optional Epi-UV illumination unit can also be installed. Thus the system can be perfectly adapted to experimental conditions.
Ordering information: The powerful BIO-VISION software (included) offers extensive possibilities of recording, display, analysis and output, and is clearly designed and easy to use.
4 Options Available Below
Product Details & Documents
The Quantum CX5 Edge gel documentation system is an ultra-modern, fully automated, yet compact system with innovative features and superior technology. With just one click, it allows the recording of images of the highest quality ('One-Button-Imaging').
- Easy to operate, standalone
- 'Scientific-grade' camera with 5 MP resolution and 6× automated zoom lens
- Robust and light-proof all-metal darkroom
- Including extendable transilluminator (optional as UV, SUPER-BRIGHT, blue and white light) usable with only one hand and interchangeable
- Unprecedented sensitivity on a receiving surface of up to 21×26 cm
For maximum flexibility in fluorescence applications an optional Epi-UV illumination unit can also be installed. Thus the system can be perfectly adapted to experimental conditions.
Ordering information: The powerful BIO-VISION software (included) offers extensive possibilities of recording, display, analysis and output, and is clearly designed and easy to use.
Product Family Options
Product Information
- AvailabilityPrice
- VILB101196701Gel documentation system, Quantum CX5 Edge, with Epi UV for TLCGel documentation system, Quantum CX5 Edge, with Epi UV for TLCLog in for availability1 * 1 ST10 550,00 €
- VILB101196791Gel documentation system, Quantum CX5 Edge, with Epi UV PadBoxGel documentation system, Quantum CX5 Edge, with Epi UV PadBoxLog in for availability1 * 1 ST9 890,00 €
- VILB101176791Gel documentation system, Quantum CX5 Edge, with Epi UV, blue
- VILB101156791Gel documentation system, Quantum CX5 Edge, with Padbox
Accessories
Product Information
- Catalog #AvailabilityPrice
- Power cord with EU plug