Size exclusion chromatography columns, PL aquagel-OH™ analytical
Supplier: VARIAN
Certificates
About this item
These PL aquagel-OH™ analytical coluns have a highly stable matrix ensuring reliable separations, even with modified eluents.
- Highly versatile for neutral, polar, anionic and cationic samples
- Mixed columns cover a wide range of molecular weights, simplifying column selection
These series have a pH range of 2-10, compatibility with organic solvent up to 50% methanol, mechanical stability up to 140 bar and low column operating pressures.