Benzopurpurine 4B, powder
Catalog # ICNA0219506925
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:724.72 g/mol
Melting Pt:554 °F (dec.)
EINECS:213-594-3
:GHS08
Storage Temperature:Ambient
CAS Number:992-59-6
Formula:C₃₄H₂₈N₆O₆S₂
MDL Number:MFCD00012410