Order Entry
Germany
ContactUsLinkComponent
 
Streptococcus (Lancefield group C strain) streptokinase, MP Biomedicals
 
Streptococcus (Lancefield group C strain) streptokinase, MP Biomedicals

 

  • CAS No.:
    9002-01-1
  • Species:
    Streptococcus (Lancefield group C strain)
  • Storage conditions:
    −20 °C
  • Form:
    White lyophilized powder
  • Enzyme name:
    Streptokinase
  • Enzyme activity:
    ≥5000 units/mg dry weight
  • Molecular weight:
    47,4 kDa