ContactUsLinkComponent
Vitamin mixture, AIN-93-VX, MP Biomedicals
Vitamin mixture, AIN-93-VX, MP Biomedicals
Catalog # ICNA0296040201
Supplier:  MP Biomedicals
undefined
Vitamin mixture, AIN-93-VX, MP Biomedicals
Catalog # ICNA0296040201
Supplier:  MP Biomedicals
Specifications
  • Description:
    AIN-93-VX vitamin mixture
  • Pk:
    1 kg
Specifications
About this item
AIN-93-VX vitamin mixture is a component of AIN-93M and AIN-93G diet.
This vitamin mix is used for the rodents diet revised after numerous nutritional and technical problems were encountered, especially during growth, pregnancy, and lactation.
Caution: Not available in pellet form.