Soy Flour, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Soy flour or soy grits refers to defatted soybeans ground finely.
- Crude Protein > 48%, Crude Fat > 0.5%, Crude Fibre < 3.5%
- Used in animal research diet
Soy flour or soy grits refers to defatted soybeans ground finely.