Vitamin Diet Fortification Powder, MP Biomedicals
Catalog # ICNA0290465401
Supplier: MP Biomedicals
Specifications
- Description:Vitamin diet fortification powder
- Pk:1 kg
Specifications
About this item
Vitamin diet fortification mixture formulation provides sufficient vitamin levels to prevent deficiencies in all animal species, giving precise control of vitamin levels.
The ingredients of the vitamin diet fortification mixture are triturated in dextrose (1 kg is sufficient for 100 lbs of diet).