Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Corn oil
Corn oil (from corn), yellow oil
Corn oil (from corn), yellow oil
Corn oil (from corn), yellow oil
Supplier:
MP Biomedicals
Certificates
Expand image
Corn oil (from corn), yellow oil
Supplier:
MP Biomedicals
Certificates
About this item
CAS Number:
8001-30-7
:
MDL Number:
MFCD00130865
Storage Temperature:
Ambient
Specification Test Results
Identity Test:
Passes
Specification Test Results
Related Information
Documentation
Certificates
Search by Lot Number
Documentation