Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Coconut oil
Coconut oil, semi-solid, hydrogenated
Coconut oil, semi-solid, hydrogenated
Coconut oil, semi-solid, hydrogenated
Supplier:
MP Biomedicals
Certificates
Expand image
Coconut oil, semi-solid, hydrogenated
Supplier:
MP Biomedicals
Certificates
About this item
CAS Number:
8001-31-8
:
MDL Number:
MFCD00130817
Melting Pt:
77.0 °CF (lit.)
Storage Temperature:
Ambient
Specification Test Results
Identity Test:
Passes
Specification Test Results
Related Information
Documentation
Certificates
Search by Lot Number
Documentation